missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse SNCA Protein
A cDNA sequence encoding the SNCA was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13535-1mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
SNCA Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MDVFMKGLSK AKEGVVAAAE KTKQGVAEAA GKTKEGVLYV GSKTKEGVVH GVTTVAEKTK EQVTNVGGAV VTGVTAVAQK TVEGAGNIAA ATGFVKKDQM GKGEEGYPQE GILEDMPVDP GSEAYEMPSE EGYQDYEPEA | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
Untagged | |
The SNCA protein contains 20mM Tris-HCl buffer (pH 7.5) and 10% glycerol. |