missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Twisted Gastrulation (TSG) Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Marke:  R&D Systems™ 756-TG-050/CF

Weitere Details : Netto-Gewicht : 0.00970kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 18347487

  • 433.03 € / 50 Mikrogramm
Verfügbar ab: 05-06-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

The Recombinant Mouse Twisted Gastrulation (TSG) Protein is derived from NS0. The Recombinant Mouse Twisted Gastrulation (TSG) Protein has been validated for the following applications: Bioactivity.
Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS.
23.5 kDa
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
65960
TSG
Mouse myeloma cell line,NS0-derived mouse TSG protein Cys25-Phe222,with a C-terminal 10-His tag
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote