missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Twisted Gastrulation (TSG) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Marke: R&D Systems™ 756-TG-050
Weitere Details : Netto-Gewicht : 0.00970kg
Beschreibung
The Recombinant Mouse Twisted Gastrulation (TSG) Protein is derived from NS0. The Recombinant Mouse Twisted Gastrulation (TSG) Protein has been validated for the following applications: Bioactivity.Spezifikation
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | |
23.5 kDa | |
50μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
65960 | |
TSG | |
Mouse myeloma cell line,NS0-derived mouse TSG protein Cys25-Phe222,with a C-terminal 10-His tag | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |