missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Der F1 Protein
A cDNA sequence encoding the Der F1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP11656-100ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
Der F1 Protein | |
Research Use Only | |
DERF1 | |
Other | |
His | |
60mM NaCl, 50mM Tris-HCl pH 8.0 and 1.2M Urea. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MKFVLAIASL LVLSTVYARP ASIKTFEEFK KAFNKNYATV EEEEVARKNF LESLKYVEAN KGAINHLSDL SLDEFKNRYL MSAEAFEQLK TQFDLNAETS ACRINSVNVP SELDLRSLRT VTPIRMQGGC GSCWAFSGVA ATESAYLAYR NTSLDLSEQE LVDCASQHGC HGDTIPRGIE YIQQNGVVEE RSYPYVAREQ RCRRPNSQHY GISNYCQIYP PDVKQIREAL TQTHTAIAVI IGIKDLRAFQ HYDGRTIIQH DNGYQPNYHA VNIVGYGSTQ GDDYWIVRNS WDTTWGDSGY GYFQAGNNLM MIEQYPYVVI M | |
Protein is >95% pure as determined by 10% PAGE (coomassie staining) and RP-HPLC. |