missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Protease Protein
A cDNA sequence encoding the Protease was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13159-150IU
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
Protease Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli |
150 IU | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged |