Learn More
enQuireBio™ Recombinant Porcine IL 1 beta Protein
A cDNA sequence encoding the IL 1 beta was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP10687-10ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
397122 | |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50 as determined by a cell proliferation assay using murine D10S cells is less than 5.0 ng/ml, corresponding to a specific activity of > 2.0 M 105 IU/mg. | |
Porcine | |
Untagged | |
The IL 1 beta was lyophilized from a 0.2microm filtered concentrated solution in PBS pH 7.4, containing 3 % trehalose. |
IL 1 beta Protein | |
Research Use Only | |
IL1B | |
Recombinant Protein | |
E. coli | |
ANVQSMECKL QDKDHKSLVL AGPHMLKALH LLTGDLKREV VFCMSFVQGD DSNNKIPVTL GIKGKNLYLS CVMKDNTPTL QLEDIDPKRY PKRDMEKRFV FYKTEIKNRV EFESALYPNW YISTSQAEQK PVFLGNSKGR QDITDFTMEV LSP | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |