missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IL 17E Protein
A cDNA sequence encoding the IL 17E was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12393-5ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
IL 17E Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
CSHLPRCCPS KQQEFPEEWL KWNPAPVSPP EPLRHTHHPE SCRASKDGPL NSRAISPWSY ELDRDLNRVP QDLYHARCLC PHCVSLQTGS HMDPMGNSVP LYHNQTVFYR RPCHGEQGAH GRYCLERRLY RVSLACVCVR PRMMA | |
Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Rat | |
Untagged | |
Lyophilized from a concentrated (1 mg/ml) solution containing no additives. |