missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Thyroxine 5-deiodinase Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Thyroxine 5-deiodinase, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP7221-ye-100ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_058906.3 | |
Thyroxine 5-deiodinase Protein | |
Research Use Only | |
Dio3 | |
Recombinant Protein | |
Yeast | |
DFLCIRKHFL RRRHPDHPEP EVELNSEGEE MPPDDPPICV SDDNRLCTLA SLKAVWHGQK LDFFKQAHEG GPAPNSEVVR PDGFQSQRIL DYAQGTRPLV LNFGSCTUPP FMARMSAFQR LVTKYQRDVD FLIIYIEEAH PSDGWVTTDS PYVIPQHRSL EDRVSAARVL QQGAPGCALV LDTMANSSSS AYGAYFERLY VIQSGTIMYQ GGRGPDGYQV SELRTWLERY DEQLHGTRPR RL | |
Greater than 90% as determined by SDS-PAGE. |
29475 | |
100 μg | |
Not determined. | |
Extracellular Domain | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |