Learn More
enQuireBio™ Recombinant S. aureus Escherichia coli ssPA / Stringent starvation Protein
Recombinant Protein
Marke: enQuireBio™ QP5524-5mg
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
WP_000676548.1 | |
5 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MLPNNDRHQI TDTTNGHYAP VTYIQVEAPT GTFIASGVVV GKDTLLTNKH VVDATHGDPH ALKAFPSAIN QDNYPNGGFT AEQITKYSGE GDLAIVKFSP NEQNKHIGEV VKPATMSNNA ETQVNQNITV TGYPGDKPVA TMWESKGKIT YLKGEAMQYD LSTTGGNSGS PVFNEKNEVI GIHWGGVPNE FNGAVFINEN VRNFLKQNIE DIHFANDDQP NNPDNPDNPN NPDNPNNPDE PNNPDNPNNP DNPDNGDNNN SDNPDAA | |
Greater than 95% as determined by SDS-PAGE. |
S. aureus Escherichia coli ssPA / Stringent starvation Protein | |
Research Use Only | |
ssPA | |
S. aureus | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |