missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. cerevisiae DNA repair and recombination protein RAD52 Protein

A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) DNA repair and recombination protein RAD52, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP7366-ec-1mg

Detalles adicionales : Netto-Gewicht : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16088470

  • 3405.00 € / 1 Milligramm
Verfügbar ab: 31-01-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Descripción

Descripción

Especificaciones

Especificaciones

NP_013680.2
S. cerevisiae DNA repair and recombination protein RAD52 Protein
Research Use Only
RAD52
Recombinant Protein
E. coli
IFGYNGWSTE VKSVVIDFLD ERQGKFSIGC TAIVRVTLTS GTYREDIGYG TVENERRKPA AFERAKKSAV TDALKRSLRG FGNALGNCLY DKDFLAKIDK VKFDPPDFDE NNLFRPTDEI SESSRTNTLH ENQEQQQYPN KRRQLTKVTN TNPDSTKNLV KIENTVSRGT PMMAAPAEAN SKNSSNKDTD LKSLDASKQD QDDLLDDSLM FSDDFQDDDL INM
Greater than 90% as determined by SDS-PAGE.
854976
1 mg
Not determined.
Partial (see sequence information for more details).
S. cerevisiae
His-SUMO
Tris-based buffer, 50% glycerol
Videos
SDS
Documentos

Documentos

Product Certifications
Ofertas especiales

Ofertas especiales