missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Soybean Soybean P34 Protein
A cDNA sequence encoding the Soybean P34 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13563-100ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
P34 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
50mM Tris-HCl, pH 8.0 and 60mM NaCl. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Soybean | |
His | |
Protein is >95% pure as determined by 10% PAGE (coomassie staining) and RP-HPLC. |