Learn More
Thermo Scientific™ REG3A Recombinant Human Protein, His Tag
Expressed from DNA sequence encoding human REG3A (Met 1-Asp 175) precursor fused to polyhistidine tag at C-terminus
Marke: Thermo Scientific™ 11235H08H250
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Expressed from DNA sequence encoding human REG3A (Met 1-Asp 175) precursor fused to polyhistidine tag at C-terminus
- Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
Spezifikation
Protein Structure Analysis, Immunogen, Protein Standard, Cell Biology and Analysis | |
18kd | |
Human | |
<1 EU/μg | |
REG3A | |
His Tag | |
Other proteins | |
≥95% by SDS-PAGE |
5068 | |
5 x 50μg | |
Full Length, Recombinant | |
Regenerating Islet-Derived Protein 3-Alpha | |
REG3A Recombinant Human Protein | |
Mammalian | |
Lyophilized |
For Research Use Only. Not for use in diagnostic procedures.