missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Maintenance Supplies for Capillary Columns

“Lock” capillary columns to the correct insertion distance recommended by the instrument manufacturer during installation. Restek™ Slide-Lock Tweezers and 15cm Compact Steel Ruler are useful for many laboratory tasks.

Marke:  Restek™ 20101

Weitere Details : Netto-Gewicht : 0.50000kg

Artikelnummer. 15104149

  • 43.20 € / 1 Stück
Verfügbar ab: 22-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

  • An alternative way to install capillary columns to the correct insertion distance recommended by the instrument manufacturer
  • Useful for many laboratory tasks
Spezifikation

Spezifikation

Capillary Column Installation
Capillary Columns
Steel
Slide-Lock Tweezers and 15 cm Compact Steel Ruler
15 cm
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote