Learn More
Restek™ MicroPulse Pulse Dampers
Compact unit can be placed almost anywhere
978.75 €
Spezifikation
Typ | Minipulse Pulse Dampener |
---|---|
Zur Verwendung mit (Anwendung) | For Applications Where Minimizing The Total System Volume is Critical |
Volumen (metrisch) | 160 μL |
Beschreibung
The MiniPulse pulse damper is ideal for applications where minimizing the total system volume is critical.
- Compact unit
- Small
- 160μL dead volume at atmospheric pressure
- Diameter: 2.5 x 1.5 in.
Spezifikation
Minipulse Pulse Dampener | |
For Applications Where Minimizing The Total System Volume is Critical | |
160 μL |