Learn More
Restek™ Replacement Split/Splitless Weldment
Artikelnummer. 13107142
Encompass_Preferred
Ansicht ändern
Click to view available options
Material:
Edelstahl
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Material
13107142 Edelstahl
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 13107142 Lieferant Restek™ Lieferanten-Nr. 22686
 Eingeschränkter Artikel
Verfügbar ab: 17-11-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
Direct replacement for Agilent 5890 and 6890/6850 GCs
  • Siltek™ treatment passivates the metal surface ensuring an inert pathway for the sample, delivering increased performance
  • High-quality stainless steel injector
  • Meets or exceeds Agilent original equipment performance
Spezifikation
Zur Verwendung mit (Geräte) Large Canister Type Filter, Agilent 6890/6850 Gcs
Material Edelstahl
Produkttyp Split/Splitless Weldment