Learn More
Restek™ Tube and Pipe Fittings for GC Equipment
Ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
Marke:  Restek™ 21810
Weitere Details : Netto-Gewicht : 0.25000kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 10085281
  • 180.00 € / 20 Stück
Verfügbar ab: 24-10-2023
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
  • Available in brass and stainless steel
  • Siltek/Sulfinert treated or Silcosteel-CR treated fittings
Note
Nuts and ferrules are not treated unless requested. Nuts and ferrules normally are not in contact with sample pathway, and thus do not require coating.
Spezifikation
Spezifikation
GC Equipment
Brass
20/Pk.
0.125 in.
Nut and Ferrule Set
Nuts, Back Ferrules, and Front Ferrules