Learn More
Invitrogen™ RGS6 Recombinant Rabbit Monoclonal Antibody (24GB3885)
Rabbit Recombinant Monoclonal Antibody
Marke:  Invitrogen™ MA557217
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 30275167
  • 514.00 € / 100 Mikroliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
RGS6 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
Members of the RGS (regulator of G protein signaling) family, such as RGS6, modulate G protein function by activating the intrinsic GTPase activity of the alpha (guanine nucleotide-binding) subunits (Seki et al., 1999 [PubMed 10083744]).
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
RGS6
Recombinant Monoclonal
Unconjugated
RGS6
DKFZp313G1241; FLJ43552; GAP; H_DJ1108A12.1; MGC142132; regulator of G-protein signaling 6; regulator of G-protein signalling 6; RGS6; S914; WUGSC:H_DJ0283M22.1; WUGSC:H_DJ1108A12.1
Rabbit
Affinity chromatography
RUO
50779, 54295, 9628
-20°C
Liquid
Western Blot, Immunocytochemistry
24GB3885
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P49758, P49801, Q9Z2H2
RGS6
A synthesized peptide derived from human RGS6 (aa 403-465).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen