Learn More
MedChemExpress RIPA Lysis Buffer (Strong)
Artikelnummer. 16448946
Encompass_Preferred
Change view
Click to view available options
:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer.
16448946 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 16448946 Lieferant MedChemExpress Lieferanten-Nr. HYK1001
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
MedChemExpress MCE RIPA Lysis Buffer is one of the most reliable buffers used to lyse cells from both cultured cells and tissues.
Spezifikation
Menge 100 mL
Produkttyp Kits
Inhalt und Lagerung Stored at -20°C, and is stable for up to 12 months.
Versandbedingung Room temperature
Ziel VD/VDR