missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 17810-1-AP-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The protein encoded by this gene contains a RING finger motif and acts as a transcription regulator. This protein has been shown to interact with, and inhibit the activity of, TRPS1, a transcription suppressor of GATA-mediated transcription. Transcription repressor ZNF278/PATZ is found to interact with this protein, and thus reduce the enhancement of androgen receptor-dependent transcription mediated by this protein. Studies of the mouse and rat counterparts suggested a role of this protein in spermatogenesis. A pseudogene of this gene is found on chromosome 1.Spezifikation
| RNF4 | |
| Polyclonal | |
| Unconjugated | |
| RNF4 | |
| E3 ubiquitin ligase RNF4, Protein SNURF, RES4 26, ring finger protein 4, RNF4, SNURF | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19822, 29274, 6047 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.32 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O88846, P78317, Q9QZS2 | |
| RNF4 | |
| RNF4 Fusion Protein Ag12115 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur