Learn More
STEMCELL Technologies RoboSep™ HLA Chimerism Buffy Coat CD14 Positive Selection Kit
Artikelnummer. 17117791
Encompass
Click to view available options
:
30 mL buffy coat
Artikelnummer. 17117791
Marke: STEMCELL Technologies 17878RF
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
EasySep™ kit with RoboSep™ buffer and RoboSep™ filter tips
  • The EasySep™ HLA Chimerism Buffy Coat CD14 Positive Selection Kit is designed to isolate CD14+ cells from freshly prepared buffy coat by positive selection
  • Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD14 and dextran-coated magnetic particles
  • Labeled cells are separated using an EasySep™ magnet without the use of columns
  • Cells of interest remain in the tube while unwanted cells are poured off
  • The CD14 antigen is strongly expressed on monocytes and is weakly expressed on granulocytes
  • It is also expressed on most tissue macrophages
  • This product replaces the EasySep™ Human Buffy Coat CD14 Positive Selection Kit (Catalog #18088) for even faster cell isolations
Spezifikation
Zelltyp Monocytes
Zur Verwendung mit (Anwendung) Cell Isolation
Menge 30 mL buffy coat