Learn More
STEMCELL Technologies RoboSep™ HLA Chimerism Whole Blood CD3 Positive Selection Kit
Artikelnummer. 17107781
Encompass
Click to view available options
Menge:
60 ml Buffy-Coat oder Vollblut
Packungsgröße:
1 Set
Artikelnummer. 17107781
Marke: STEMCELL Technologies 17871RF
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
EasySep™ kit with RoboSep™ buffer and RoboSep™ filter tips
  • The EasySep™ HLA Chimerism Whole Blood CD3 Positive Selection Kit is designed to isolate CD3+ cells from fresh whole blood or buffy coat by positive selection
  • Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD3 and dextran-coated magnetic particles
  • The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding
  • Labeled cells are separated using an EasySep™ magnet without the use of columns
  • Cells of interest remain in the tube while unwanted cells are poured off
  • The CD3 antigen is expressed on T cells and NKT cells
  • This product replaces the EasySep™ Human Whole Blood CD3 Positive Selection Kit (Catalog #18081) and EasySep™ HLA Whole Blood CD3 Positive Selection Kit (Catalog #18081HLA), for even faster cell isolations
Spezifikation
Zelltyp T Cells
Menge 60 ml Buffy-Coat oder Vollblut