Learn More
STEMCELL Technologies RoboSep™ Human 'Do-It-Yourself' Positive Selection Kit II
Click to view available options
Menge:
1 x 109 Zellen
Packungsgröße:
1 Set
Beschreibung
- The EasySep™ Human 'Do-It-Yourself' Positive Selection Kit II is designed to select any human cell type of interest with your own mouse IgG1 monoclonal antibody
- Cells that express the surface antigen that is recognized by the mouse IgG1 monoclonal antibody are targeted for magnetic separation using EasySep™ Dextran RapidSpheres™ and the EasySep™ Magnet
- The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding
- This product replaces the EasySep™ Human 'Do-It-Yourself' Selection Kit (Catalog #18099)
Spezifikation
Spezifikation
| Zelltyp | B Cells, Dendritic Cells, Granulocytes and Subsets, Hematopoietic Stem and Progenitor Cells, Macrophages, Marrow Stromal Cells, Mesenchymal Stem and Progenitor Cells, Monocytes, Mononuclear Cells, Myeloid Cells, NK Cells, Other, Plasma, T Cells |
| Zur Verwendung mit (Anwendung) | Cell Isolation |
| Menge | 1 x 109 Zellen |