Learn More
STEMCELL Technologies RoboSep™ Human Gamma/Delta T Cell Isolation Kit
Artikelnummer. 17197911
Encompass
Click to view available options
Menge:
1 x 109 Zellen
Packungsgröße:
1 Set
Artikelnummer. 17197911
Marke: STEMCELL Technologies 19255RF
om dit product te kopen Registreer vandaag om een webaccount aan te maken
EasySep™ kit with RoboSep™ buffer and RoboSep™ filter tips
  • The EasySep™ Human Gamma/Delta T Cell Isolation Kit is designed to isolate gamma/delta T cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection
  • Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-gamma/delta T cells and dextran-coated magnetic particles
  • The labeled cells are separated using the EasySep™ magnet without the use of columns
  • The desired cells are poured off into a new tube
Spezifikation
Zelltyp T Cells, T Cells, Other Subsets
Zur Verwendung mit (Anwendung) Cell Isolation
Menge 1 x 109 Zellen