Learn More
STEMCELL Technologies RoboSep™ Human Neutrophil Isolation Kit
Artikelnummer. 17117831
Encompass
Change view
Click to view available options
:
1 x 109 cells
Artikelnummer.
17117831 1 x 109 cells
1 options
Artikelnummer. 17117831
Marke: STEMCELL Technologies 17957RF
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Immunomagnetic isolation of untouched human neutrophils
  • The EasySep™ Human Neutrophil Isolation Kit is designed to isolate neutrophils from a polymorphonuclear cell-rich fraction of peripheral blood by negative selection
  • Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-neutrophils and magnetic particles
  • Labeled cells are separated using an EasySep™ magnet without the use of columns
  • Desired cells are poured off into a new tube
  • The isolation kit is compatible with cells prepared using HetaSep™ (Catalog #07906) sedimentation or red blood cell lysis
  • This product replaces the EasySep™ Human Neutrophil Enrichment Kit (Catalog #19257), for even faster cell isolations
Spezifikation
Zelltyp Granulocytes and Subsets
Zur Verwendung mit (Anwendung) Cell Isolation
Menge 1 x 109 cells