Learn More
STEMCELL Technologies RosetteSep™ Human B Cell Enrichment Cocktail
Beschreibung
- The RosetteSep™ Human B Cell Enrichment Cocktail is designed to isolate B cells from whole blood by negative selection
- Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-B cells and glycophorin A on red blood cells (RBCs)
- When centrifuged over a buoyant density medium such as RosetteSep™ DM-L (Catalog #15705) or Lymphoprep™ (Catalog #07801), the unwanted cells pellet along with the RBCs
- The purified B cells are present as a highly enriched population at the interface between the plasma and the buoyant density medium
Spezifikation
Spezifikation
| Zelltyp | B Cells |
| Zur Verwendung mit (Anwendung) | Cell Isolation |
| Menge | 200 mL Vollblut |