Learn More
STEMCELL Technologies RosetteSep™ Human Bone Marrow Progenitor Cell Pre-Enrichment Cocktail
Artikelnummer. 17127671
Encompass
Change view
Click to view available options
Menge:
200 ml Knochenmark
40 mL Knochenmark
Packungsgröße:
1 Set
Artikelnummer. Menge unitSize
17127671 200 ml Knochenmark 1 Set
17157661 40 mL Knochenmark 1 Set
2 options
Artikelnummer. 17127671
Marke: STEMCELL Technologies 15067
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Immunodensity pre-enrichment of untouched stem/progenitor cells from bone marrow
  • The RosetteSep™ Human Bone Marrow Progenitor Cell Pre-Enrichment Cocktail is designed to pre-enrich progenitor cells by removing lineage-specific cells from whole bone marrow by negative selection
  • Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD3, CD11b, CD14, CD16, CD19, CD56, CD66b and glycophorin A on red blood cells (RBCs)
  • When centrifuged over a buoyant density medium such as Lymphoprep™ (Catalog #07801), the unwanted cells pellet along with the RBCs
  • The pre-enriched progenitor cells are present as an enriched population at the interface between the plasma and the buoyant density medium
Spezifikation
Zelltyp Hematopoietic Stem and Progenitor Cells
Zur Verwendung mit (Anwendung) Cell Isolation
Menge 200 ml Knochenmark