Learn More
Carl Roth™ Rotilabo™ Transparent Polypropylene Female Luer Tubing Connectors
Marke: Carl Roth™ CT62.1
Weitere Details : Netto-Gewicht : 0.10000kg
Spezifikation
Female Luer Tubing Connector | |
Polypropylene | |
10 Pack |
Autoclavable | |
1.6mm I.D. Hoses | |
Transparent |