Learn More
Invitrogen™ RPGR Recombinant Rabbit Monoclonal Antibody (0S8U8)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA558011
Beschreibung
Immunogen sequence: MREPEELMPD SGAVFTFGKS KFAENNPGKF WFKNDVPVHL SCGDEHSAVV TGNNKLYMFG SNNWGQLGLG SKSAISKPTC VKALKPEKVK LAACGRNHTL VSTEGGNVYA TGGNNEGQLG LGDTEERNTF HVISFFTSEH KIKQLSAGSN TSAALTEDGR LFMWGDNSEG QIGLKNVSNV CVPQQVTIGK PVSWISCGYY HSAFVTTDGE LYVFGEPENG KLGLPNQLLG NHRTPQLVSE IPEKVIQVAC GGEHTVVLTE.
This gene encodes a protein with a series of six RCC1-like domains (RLDs), characteristic of the highly conserved guanine nucleotide exchange factors. The encoded protein is found in the Golgi body and interacts with RPGRIP1. This protein localizes to the outer segment of rod photoreceptors and is essential for their viability. Mutations in this gene have been associated with X-linked retinitis pigmentosa (XLRP). Multiple alternatively spliced transcript variants that encode different isoforms of this gene have been reported, but the full-length natures of only some have been determined.
Spezifikation
| RPGR | |
| Recombinant Monoclonal | |
| 1.49 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q9R0X5 | |
| RPGR | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human RPGR (NP_001030025.1). | |
| 20 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 0S8U8 | |
| Unconjugated | |
| RPGR | |
| COD1; CORDX1; CRD; mRpgr; orf15; PCDX; retinitis pigmentosa 15; retinitis pigmentosa 3 GTPase regulator; retinitis pigmentosa GTPase regulator; RP15; RP3; Rp3h; Rpgr; X-linked retinitis pigmentosa GTPase regulator; XLRP3 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 19893, 367733 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.