missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS3 Mouse anti-Human, Mouse, Rat, Clone: 2G7H4, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66046-1-IG-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
WD repeat domain 5 (WDR5) is a member of the WD repeat protein family, which is involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. WDR5, also known as BIG-3, is expressed in the developing growth plate, accelerates chondrocyte and osteoblast differentiation in vitro, and regulates osteoblast differentiation during embryonic bone development. WDR5 interacts with the pluripotency factor Oct4/POU5F1 and is required for the efficient formation of induced pluripotent stem (iPS) cells.Spezifikation
RPS3 | |
Monoclonal | |
1.12 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P23396, P62908, P62909 | |
RPS3 | |
RPS3 Fusion Protein Ag7515 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG2a |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
2G7H4 | |
Unconjugated | |
RPS3 | |
40S ribosomal protein S3, ribosomal protein S3, RPS3 | |
Mouse | |
Protein A | |
RUO | |
140654, 27050, 6188 | |
-20°C | |
Liquid |