missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RRP1B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 27723-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
RRP1B belongs to the RRP1 family. It may be a novel susceptibility gene for breast cancer progression and metastasis.Spezifikation
RRP1B | |
Polyclonal | |
Unconjugated | |
RRP1B | |
KIAA0179, Nnp1, NNP1L, RRP1, RRP1 like protein B, RRP1B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23076 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunocytochemistry | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q14684 | |
RRP1B | |
RRP1B Fusion Protein Ag23405 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |