missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-((tert-Butoxycarbonyl)amino)-5-methoxy-5-oxopentanoic acid, 97%, BLD Pharm™

Artikelnummer. 30987687
Encompass_Preferred
Ansicht ändern
Click to view available options
Menge:
5 g
25 g
100 g
500 g
1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
30987687 500 g
30969378 25 g
30984762 100 g
30982668 1 kg
30979214 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30987687 Lieferant BLD Pharm Lieferanten-Nr. BD118097500g
 Eingeschränkter Artikel
Verfügbar ab: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Chemischer Name oder Material (S)-2-((tert-Butoxycarbonyl)amino)-5-methoxy-5-oxopentanoic acid
CAS 45214-91-3
Summenformel C11H19NO6
MDL-Nummer MFCD00190794
Menge 500 g
SMILES O=C(OC)CC[C@@H](C(O)=O)NC(OC(C)(C)C)=O
Molekulargewicht (g/mol) 261.27
Reinheit (%) 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.