missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-Ethyl-4,9-dihydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, 98%, BLD Pharm™

Product Code. 31073872
Encompass_Preferred
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Menge:
100 mg
250 mg
1 g
5 g
25 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge
31073872 100 mg
31063727 25 g
31072566 250 mg
31062359 5 g
31065244 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 31073872 missing translation for 'mfr' BLD Pharm missing translation for 'supplierNo' BD1153100mg
 Eingeschränkter Artikel
Verfügbar ab: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Chemischer Name oder Material (S)-4-Ethyl-4,9-dihydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione
CAS 19685-09-7
Summenformel C20H16N2O5
MDL-Nummer MFCD02093100
Menge 100 mg
SMILES O=C1[C@](O)(CC)C2=C(CO1)C(N3CC4=CC5=CC(O)=CC=C5N=C4C3=C2)=O
Molekulargewicht (g/mol) 364.35
Reinheit (%) 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.