missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A11 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA521330
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: A431. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in motility, invasion, and tubulin polymerization. Chromosomal rearrangements and altered expression of this gene have been implicated in tumor metastasis.Spezifikation
S100A11 | |
Polyclonal | |
Unconjugated | |
S100A11 | |
cal; Calgizzarin; calizzarin; EMAP; Emap1; EMAPI; endothelial monocyte activating polypeptide 1; endothelial monocyte-activating polypeptide; epididymis secretory protein Li 43; HEL-S-43; Metastatic lymph node gene 70 protein; MLN 70; MLN70; Protein S100-A11; Protein S100-A11, N-terminally processed; Protein S100C; Protein S100-C; S100 calcium binding protein A11; S100 calcium binding protein A11 (calgizzarin); S100 calcium binding protein A11 (calizzarin); S100 calcium binding protein A14; S100 calcium-binding protein A11; S100 calcium-binding protein A11 (calgizzarin); S100a11; S100a14; S100C | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6282 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P31949 | |
S100A11 | |
Synthetic peptide corresponding to a region within amino acids 8 and 102 of S100A11 (Uniprot ID#P31949) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |