Learn More
S100A14 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA555666
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: GQCRSANAED AQEFSDVERA IETLIKNFHQ YSVEGGKETL TPSELRDLVT QQLPHLMPSN CGLEEKIANL GSCNDSKLEF RSFWELIGEA Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 37%.
S100A14 gene encodes a member of the S100 protein family which contains an EF-hand motif and binds calcium. The gene is located in a cluster of S100 genes on chromosome 1. Levels of the encoded protein have been found to be lower in cancerous tissue and associated with metastasis suggesting a tumor suppressor function.Spezifikation
S100A14 | |
Polyclonal | |
Unconjugated | |
S100a14 | |
1110013O05Rik; BCMP84; breast cancer membrane protein 84; Gm1020; Protein S100-A14; S100 calcium binding protein A14; S100 calcium binding protein A15; S100 calcium-binding protein A14; S100A14; S100a15; S114 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57402 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HCY8 | |
S100a14 | |
Recombinant protein corresponding to Human S100A14 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only