missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A16 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA550098
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total S100A16 protein.
S100A16 (S100 Calcium Binding Protein A16) is a Protein Coding gene. Diseases associated with S100A16 include Robinow Syndrome, Autosomal Recessive and Robinow Syndrome. Gene Ontology (GO) annotations related to this gene include protein homodimerization activity. Calcium-binding protein. Binds one calcium ion per monomer. Can promote differentiation of adipocytes (in vitro). Overexpression in preadipocytes increases their proliferation, enhances adipogenesis and reduces insulin-stimulated glucose uptake.Spezifikation
S100A16 | |
Polyclonal | |
Unconjugated | |
S100a16 | |
2300002L21Rik; AAG13; aging-associated gene 13 protein; aging-associated protein 13; AI325039; AI663996; DT1P1A7; MGC17528; protein S100-A16; Protein S100F; Protein S100-F; S100 calcium binding protein A16; S100 calcium-binding protein A16; S100A16; S100F | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
140576, 361991, 67860 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q96FQ6, Q9D708 | |
S100a16 | |
Synthesized peptide derived from internal of human S100A16. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |