Learn More
S100A3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA542097
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: MARPLEQAVA AIVCTFQEYA GRCGDKYKLC QAELKELLQK ELATWTPTEF Sequence homology: Cow: 82%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%
S100A3 Polyclonal antibody specifically detects S100A3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
S100A3 | |
Polyclonal | |
Unconjugated | |
S100A3 | |
Protein S100-A3; Protein S-100E; S100 calcium binding protein A3; S100 calcium-binding protein A3; S100A3; S100E | |
Rabbit | |
Protein A | |
RUO | |
6274 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P33764 | |
S100A3 | |
Synthetic peptide directed towards the N-terminal of human S100A3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |