Learn More
S100G Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA563195
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: KSPEELKRIF EKYAAKEGDP DQLSKDELKL LIQAEFPSLL KGPNTLDDLF QELDK Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 80%.
S100G encodes calbindin D9K, a vitamin D-dependent calcium-binding protein. This cytosolic protein belongs to a family of calcium-binding proteins that includes calmodulin, parvalbumin, troponin C, and S100 protein. In the intestine, the protein is vitamin D-dependent and its expression correlates with calcium transport activity. The protein may increase Ca2+ absorption by buffering Ca2+ in the cytoplasm and increase ATP-dependent Ca2+ transport in duodenal basolateral membrane vesicles.Spezifikation
S100G | |
Polyclonal | |
Unconjugated | |
S100G | |
9 kDa CaBP; AU042539; CABP; CABP1; CABP9K; CaBP-D9K; Calb3; calbindin 3, (vitamin D-dependent calcium binding protein); calbindin 3, (vitamin D-dependent calcium-binding protein); calbindin D9k; calbindin-D9K; calbindin-D9K major form; calcium-binding protein, intestinal, vitamin D-dependent (9-kDa CaBP); Cbpi; cholecalcin; cytidine 5'-triphosphate synthase 2; protein S100-G; Rncalbd9; S100 calcium binding protein G; S100 calcium-binding protein G; S100D; S100g; Vitamin D-dependent calcium-binding protein, intestinal | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
795 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P29377 | |
S100G | |
Recombinant protein corresponding to Human S100G | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only