Learn More
UVP Filter square filter for GelDoc-It (included)
Marke:  UVP 38-0220-01
Weitere Details : Netto-Gewicht : 0.50000kg
Artikelnummer. 12893049
  • 373.00 € / 1 Stück
Verfügbar ab: 19-07-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
A variety of UV transilluminator models are available. Optional Doc-It™ LS 1D software loads onto your existing computer for analysis of images. The package offers many image enhancement tools, MW and mass calculation functions, reporting capabilities and supports 21 CFR Part 11 compliance. Dimensions, mm: 780 x 361 x 338.
There are two versions of the BioDoc-It TS system available:
BioDoc-It TS 210
- Includes a 0.3Mp camera and has one access door to the front.
BioDoc-It TS 220
- Includes a 1.3Mp camera and has three access doors. One to the front and one on each side (to facilitate band cutting).Both units incorporate the following features:
  • 8inch touch screen that pivots for optimum viewing angle
  • Network connection and USB port on the front for quick image transfer
  • Live preview and a manual zoom lens
  • Ethidium™ Bromide filter in a tray for quick exchange
  • Built-in bright overhead LED white light for focusing as well as an illumination source for white light or colorimetric samples
  • UV safety switch turns the UV off when the door is open
  • Connects directly to any USB digital thermal printer (or prints can be made via the network to any standard printer)
The BioDoc-It TS system is a stand-alone unit for imaging DNA and protein gels. It is very quick and easy to use; having been designed to suit the busy, multi-user lab.