Learn More
Thermo Scientific™ Samco™ Histology Bio-Tite™ Nonsterile Specimen Containers
Prevent bio-hazardous spills and conserve storage space with these stackable containers, offered in capacities from 20mL to 120mL with 35, 48 or 53mm openings.
Marke: Thermo Scientific™ 6A0008
Weitere Details : Netto-Gewicht : 0.40000kg
Beschreibung
Prevent bio-hazardous spills and conserve storage space with stackable Thermo Scientific™ Samco™ Histology Bio-Tite™ Nonsterile Specimen Containers, offered in capacities from 20mL to 120mL with 35, 48 or 53mm openings. These shatter-resistant containers feature a proprietary, all plastic, leak-resistant seal. They are tested at varying pressures, making them ideal for use in various transport systems.
Features:- Dual-threaded caps create leak-resistant seal for superior safety and handling
- Top and bottom positive grip knurls are easy to grip and open using latex gloves
- Virgin polypropylene allows higher clarity to see samples, and will not become brittle
Ordering Information:
Bulk packed, one bag per case. Caps must be ordered separately.
Spezifikation
Specimen Container | |
Cylindrical | |
20 mL | |
Bio-Tite Lid | |
35 mm | |
1000/Cs. |
Virgin Polypropylene | |
Non-sterile | |
35 mm | |
20mL/35mm Containers only | |
No Label | |
1000/Pk. |