Learn More
Novus Biologicals™ SAMD8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke: Novus Biologicals™ NBP3-30871-100ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
SAMD8 Polyclonal antibody specifically detects SAMD8 in Human samples. It is validated for ELISA,Western Blot
Spezifikation
| SAMD8 | |
| Polyclonal | |
| Western Blot : 5-10 μg/mL, ELISA | |
| FLJ25082, SAM domain-containing protein 8, SMSr, sphingomyelin synthase related, sphingomyelin synthase-related protein 1, sterile alpha motif domain containing 8, Sterile alpha motif domain-containing protein 8 | |
| Full-length recombinant SAMD8 protein | |
| 100 μg | |
| Lipid and Metabolism | |
| 142891 | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| ELISA, Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human | |
| Purified |