missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SAP97 Recombinant Rabbit Monoclonal Antibody (24GB5305)

Rabbit Recombinant Monoclonal Antibody
146.00 € - 500.00 €
Spezifikation
| Antigen | SAP97 |
|---|---|
| Klon | 24GB5305 |
| Konzentration | 0.62 mg/mL |
| Inhalt und Lagerung | -20°C |
| Anwendungen | Flow Cytometry, Western Blot, Immunocytochemistry |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
30275661
![]() |
Invitrogen™
MA557017 |
100 μL |
500.00 €
100 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
30275541
|
Invitrogen™
MA557018 |
20 μL |
146.00 €
20 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
SAP97 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes a multi-domain scaffolding protein that is required for normal development. This protein may have a role in septate junction formation, signal transduction, cell proliferation, synaptogenesis and lymphocyte activation. Several alternatively spliced transcript variants encoding different isoforms have been described for this gene, but the full-length nature of some of the variants is not known.Spezifikation
| SAP97 | |
| 0.62 mg/mL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q12959, Q62696, Q811D0 | |
| 13383, 1739, 25252 | |
| A synthesized peptide derived from human SAP97 (aa 1-44). | |
| Antibody |
| 24GB5305 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| DLG1 | |
| B130052P05Rik; discs large homolog 1; discs large homolog 1, scribble cell polarity complex component; discs large MAGUK scaffold protein 1; discs, large homolog 1; discs, large homolog 1 (Drosophila); discs, large homolog 1 (Drosophila) transcript variant 6-v1; discs, large homolog 1 (Drosophila) transcript variant 6-v2; disks large homolog 1; dJ1061C18.1.1; DKFZp761P0818; DKFZp781B0426; DLG1; Dlgh1; Drosophila discs-large tumor suppressor homologue (synapse associated protein); E-dlg/SAP97; Embryo-dlg/synapse-associated protein 97; hDlg; mKIAA4187; presynaptic protein SAP97; SAP97; SAP-97; synapse-associated protein 97 | |
| DLG1 | |
| Primary | |
| Affinity chromatography |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts