Learn More
Carl Roth™ Sarogold™ Folie
Made with a base of vinylidene chloride. Roth Carl™ Sarogold™-foil does not stick or burn.
Marke: Carl Roth™ 9068.1
Weitere Details : Netto-Gewicht : 0.50000kg
Beschreibung
- Made on the basis of vinylidene chloride
- Approved for food wrapping, with excellent protecting qualities
- Excellent for covering gel-electrophoresis plates, particularly with DNA- and RNA-sequencing gels
- Protects against direct contact with dangerous substances like radioactive isotopes and ethidium bromide
- Does not stick and is easy to unroll
- Does not burn and is self-extinguishing
- Can be used in microwaves
- Comes in tear-off box
Spezifikation
Cutter included | |
Folie | |
300 mm | |
11.5 μm |