Learn More
SARS-CoV-2 RBD Neutralizing Monoclonal Antibody (OTI1C2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
242.00 € - 540.87 €
Spezifikation
| Antigen | SARS-CoV-2 RBD Neutralizing |
|---|---|
| Klon | OTI1C2 |
| Konzentration | 1 mg/mL |
| Anwendungen | ELISA, Neutralization |
| Klassifikation | Monoclonal |
| Product Code | Brand | Menge | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Menge | Price | Quantity & Availability | |||||
17589304
|
Origene Technologies
TA814512S |
30 μL |
242.00 €
30 Mikroliter |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | |||||
17569304
|
Origene Technologies
TA814512 |
100 μL |
540.87 €
100 Mikroliter |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | |||||
Description
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
SARS-CoV-2 RBD Neutralizing Monoclonal antibody specifically detects SARS-CoV-2 RBD Neutralizing in Virus samples. It is validated for ELISA, NeutralizationSpecifications
| SARS-CoV-2 RBD Neutralizing | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Recombinant fusion protein of SARS-CoV-2 Spike RBD protein and virion surface domain of membrane protein (M-protein) produced in HEK293 | |
| Primary | |
| -20° C, Avoid Freeze/Thaw Cycles |
| OTI1C2 | |
| ELISA, Neutralization | |
| Unconjugated | |
| Mouse | |
| Virus | |
| Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2 | |
| IgG1 | |
| Protein A/G | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.