missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems SARS-CoV-2 S protein (D614G), His Tag, Super stable trimer
MALS verified
544.34 € - 3762.02 €
Spezifikation
Zugriffsnummer | QHD43416.1 (D614G) |
---|---|
Molekulargewicht | 138.0 kDa |
Quelle | SARS-CoV-2 S protein (D614G), His Tag, Super stable trimer (SPN-C52H3) is the ectodomain of SARS-CoV-2 S protein that contains AA Val 16 - Pro 1213 (Accession # QHD43416.1) and D614G mutation, which has become increasingly common in SARS-CoV-2 viruses from around the world . The recombinant protein is expressed from human 293 cells (HEK293) with T4 fibritin trimerization motif and a polyhistidine tag (at the C-terminus. Proline substitutions (F817P, A892P, A899P, A942P, K986P, V987P) and alanine substitutions (R683A and R685A) are introduced to stabilize the trimeric prefusion state of SARS-CoV-2 S protein and abolish the furin cleavage site, respectively.) |
Lagerungsbedingungen | -20°C |
Gen-Alias | Spike,S protein,Spike glycoprotein,S glycoprotein,COVID-19 |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
17106771
|
ACROBiosystems
SPN-C52H3-50UG |
50 μg |
544.34 €
50 Mikrogramm |
Verfügbar ab: 21-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
17196761
|
ACROBiosystems
SPN-C52H3-500UG |
500 μg |
3762.02 €
500 Mikrogramm |
Verfügbar ab: 21-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 138.0 kDa. The protein migrates as 170-200 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
QHD43416.1 (D614G) | |
SARS-CoV-2 S protein (D614G), His Tag, Super stable trimer (SPN-C52H3) is the ectodomain of SARS-CoV-2 S protein that contains AA Val 16 - Pro 1213 (Accession # QHD43416.1) and D614G mutation, which has become increasingly common in SARS-CoV-2 viruses from around the world . The recombinant protein is expressed from human 293 cells (HEK293) with T4 fibritin trimerization motif and a polyhistidine tag (at the C-terminus. Proline substitutions (F817P, A892P, A899P, A942P, K986P, V987P) and alanine substitutions (R683A and R685A) are introduced to stabilize the trimeric prefusion state of SARS-CoV-2 S protein and abolish the furin cleavage site, respectively.) | |
Spike,S protein,Spike glycoprotein,S glycoprotein,COVID-19 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
138.0 kDa | |
-20°C | |
S protein | |
SARS-CoV-2 | |
PBS | |
HEK293 | |
Viral Proteins |