missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS Nucleocapsid Protein Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 49 publications
Marke: Novus Biologicals NB100-56576SS
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
SARS Nucleocapsid Protein Polyclonal specifically detects SARS Nucleocapsid Protein in SARS-CoV-2, SARS-CoV, Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Dual RNAscope ISH-IHC.
Spezifikation
| SARS Nucleocapsid Protein | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| 2019-nCoV Nucleocapsid Protein, COVID-19 N protein, N, N protein, N structural protein, NC, Nucleocapsid protein, Nucleoprotein, Protein N, SARS coronavirus N protein, SARS coronavirus nucleocapsid protein, SARS CoV, SARS CoV N protein, SARS CoV nucleocapsid protein, SARS N protein, SARS Nucleoprotein, SARSCoV, SARSCoV N protein, SARSCoV nucleocapsid protein, SARS-CoV-2 N protein, Severe acute respiratory syndrome | |
| The antibody was developed by immunizing Rabbit with a synthetic peptide corresponding to amino acids 399-411 (DLDDFSKQLQQSM-C) from the N (SARS Nucleocapsid) for the Human SARS coronavirus (Genbank accession no. YP_009724397.2) | |
| 0.025 mg | |
| Primary | |
| SARS-CoV-2, SARS-CoV, Virus | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), In Situ Hybridization (ISH) | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000, Immunohistochemistry reported in scientific literature (PMID 32396922), Immunocytochemistry/Immunofluorescence 1:10-1:500. Use reported in scientific literature (Mossel et al (2008)), Immunohistochemistry-Paraffin reported in scientific literature (PMID 24725942), SDS-Page reported in scientific literature (PMID 34880383), Dual RNAscope ISH-IHC | |
| NP_006504.2 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| SARS Nucleocapsid protein. Dot Blot results using recombinant proteins for cross-reactivity testing revealed high reactivity to SARS-CoV-2 Nucleocapsid protein (NBP2-90975) and low/no reactivity towards H1N1 (NBP1-99041). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur