missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS Nucleocapsid Protein Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 49 publications
242.00 € - 513.00 €
Spezifikation
| Antigen | SARS Nucleocapsid Protein |
|---|---|
| Konzentration | 1.0 mg/mL |
| Verdünnung | Western Blot 1:100-1:2000, Immunohistochemistry reported in scientific literature (PMID 32396922), Immunocytochemistry/Immunofluorescence 1:10-1:500. Use reported in scientific literature (Mossel et al (2008)), Immunohistochemistry-Paraffin reported in scientific literature (PMID 24725942), SDS-Page reported in scientific literature (PMID 34880383), Dual RNAscope ISH-IHC |
| Anwendungen | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), In Situ Hybridization (ISH) |
| Klassifikation | Polyclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18046255
|
Novus Biologicals
NB100-56576SS |
0.025 mg |
242.00 €
25 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
18767943
|
Novus Biologicals
NB100-56576 |
513.00 €
0.10 Milligramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | ||||||
Beschreibung
SARS Nucleocapsid Protein Polyclonal specifically detects SARS Nucleocapsid Protein in SARS-CoV-2, SARS-CoV, Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Dual RNAscope ISH-IHC.Spezifikation
| SARS Nucleocapsid Protein | |
| Western Blot 1:100-1:2000, Immunohistochemistry reported in scientific literature (PMID 32396922), Immunocytochemistry/Immunofluorescence 1:10-1:500. Use reported in scientific literature (Mossel et al (2008)), Immunohistochemistry-Paraffin reported in scientific literature (PMID 24725942), SDS-Page reported in scientific literature (PMID 34880383), Dual RNAscope ISH-IHC | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| 2019-nCoV Nucleocapsid Protein, COVID-19 N protein, N, N protein, N structural protein, NC, Nucleocapsid protein, Nucleoprotein, Protein N, SARS coronavirus N protein, SARS coronavirus nucleocapsid protein, SARS CoV, SARS CoV N protein, SARS CoV nucleocapsid protein, SARS N protein, SARS Nucleoprotein, SARSCoV, SARSCoV N protein, SARSCoV nucleocapsid protein, SARS-CoV-2 N protein, Severe acute respiratory syndrome | |
| IgG | |
| Immunogen affinity purified | |
| SARS Nucleocapsid protein. Dot Blot results using recombinant proteins for cross-reactivity testing revealed high reactivity to SARS-CoV-2 Nucleocapsid protein (NBP2-90975) and low/no reactivity towards H1N1 (NBP1-99041). |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), In Situ Hybridization (ISH) | |
| Unconjugated | |
| Rabbit | |
| SARS-CoV-2, SARS-CoV, Virus | |
| NP_006504.2 | |
| The antibody was developed by immunizing Rabbit with a synthetic peptide corresponding to amino acids 399-411 (DLDDFSKQLQQSM-C) from the N (SARS Nucleocapsid) for the Human SARS coronavirus (Genbank accession no. YP_009724397.2) | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts