Learn More
SCNM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 15490-1-AP-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
SCNM1 is a zinc finger protein and putative splicing factor. In mice, Scnm1 modifies phenotypic expression of Scn8a mutations.Spezifikation
| SCNM1 | |
| Polyclonal | |
| Unconjugated | |
| SCNM1 | |
| SCNM1, sodium channel modifier 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 310662, 69269, 79005 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8K136, Q9BWG6 | |
| SCNM1 | |
| SCNM1 Fusion Protein Ag7854 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |