missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEC6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 14703-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene is a component of the exocyst complex, a multiple protein complex essential for targeting exocytic vesicles to specific docking sites on the plasma membrane. Though best characterized in yeast, the component proteins and functions of exocyst complex have been demonstrated to be highly conserved in higher eukaryotes. At least eight components of the exocyst complex, including this protein, are found to interact with the actin cytoskeletal remodeling and vesicle transport machinery. The complex is also essential for the biogenesis of epithelial cell surface polarity.Spezifikation
SEC6 | |
Polyclonal | |
Unconjugated | |
EXOC3 | |
EXOC3, exocyst complex component 3, Exocyst complex component Sec6, SEC6, SEC6L1, Sec6p | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11336, 211446, 252881 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O60645, Q62825, Q6KAR6 | |
Exoc3 | |
SEC6 Fusion Protein Ag6383 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |