Learn More
Invitrogen™ SEC61alpha Recombinant Rabbit Monoclonal Antibody (24GB1230)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA557248
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
SEC61alpha Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The SEC61 complex is a major component of the endoplasmic reticulum (ER) translocation site. This complex is composed of three subunits: SEC61alpha, SEC61beta and SEC61gamma. SEC61alpha is a polytopic integral membrane protein. It is a core component of the ER translocation complex, playing a crucial role in the insertion of secretory and membrane polypeptides into the ER.
Spezifikation
| SEC61alpha | |
| Recombinant Monoclonal | |
| Unconjugated | |
| SEC61A2 | |
| protein transport protein Sec61 subunit alpha; protein transport protein Sec61 subunit alpha isoform 2; Sec61 alpha; Sec61 alpha 2 subunit; Sec61 alpha 2 subunit (S. cerevisiae); Sec61 alpha form 2; sec61 alpha-2; Sec61 translocon alpha 2 subunit; Sec61, alpha subunit 2; Sec61, alpha subunit 2 (S. cerevisiae); SEC61A2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 361273, 55176, 57743 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1230 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9H9S3, Q9JLR1 | |
| SEC61A2 | |
| A synthesized peptide derived from human SEC61A1/2 (aa 350-450). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |