missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHANK1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke: Novus Biologicals NBP2-33971
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
SHANK1 Polyclonal specifically detects SHANK1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Spezifikation
| SHANK1 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Q9Y566 | |
| SHANK1 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LRSKSMTSELEEMEYEQQPAPVPSMEKKRTVYQMALNKLDEILAAAQQTISASESPGPGGLASLGKHRPKGFFATESSFDPHHR | |
| 0.1 mL | |
| Neuroscience | |
| 50944 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| SH3 and multiple ankyrin repeat domains 1, SH3 and multiple ankyrin repeat domains protein 1, Shank1, Somatostatin receptor-interacting protein, SSTR-interacting protein, SSTRIPSPANK-1, synamon | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur